Hamilton™ CO-RE™ II Conductive Filter Tips, non-sterile, 1000 μL
Thermo Scientific™
Hamilton™ CO-RE™ II Conductive Filter Tips, non-sterile, 1000 μL
These filter tips are for use on the HID NIMBUS Presto System.Read more
Have Questions?
Catalog NumberQuantity
A476193840 Tips/Cs.
Catalog number A47619
Price (USD)
432.00
Each
Quantity:
3840 Tips/Cs.
Price (USD)
432.00
Each
These filter tips are for use on the HID NIMBUS Presto System.
Specifications
Certifications/ComplianceCertified for Biological Purity as DNase/RNase free and non-pyrogenic
DisposableSingle-use
Length (Metric)95.1 mm
Quantity3840 Tips/Cs.
AutoclavableNot Autoclavable
ColorBlack
FilteredFiltered
Fit for PurposeForensic Ready
FormatBlister Refill
MaterialPolypropylene
Product LineHamilton CO-RE II
SterilityNon-sterile
Volume (Metric)1000 μL
Unit SizeEach