What is the concentration of the MS2 phage provided in the TaqMan SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit (Cat. No. A47702)?
Unfortunately, we cannot disclose the concentration of the MS2 phage in the above kit, as this information is proprietary.