Sodium Hydroxide (NaOH) Solutions, 0.1–1.0 M
Gibco™

Sodium Hydroxide (NaOH) Solutions, 0.1–1.0 M

Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentration
A4782602200 L0.1 M
A478260120 L0.1 M
A478280120 L0.5 M
A4782802200 L0.5 M
A478290120 L1.0 M
A4782902200 L1.0 M
Catalog number A4782602
Price (USD)
-
Quantity:
200 L
Concentration:
0.1 M

Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing. Available in three different grades and two off-the-shelf sizes, these solutions provide outsourcing efficiencies to help reclaim time, space, and other core resources.

Features of Gibco NaOH solutions include:

  • 0.1 M, 0.5 M, and 1 M concentrations
  • Multi-compendial grade raw materials
  • cGMP manufacturing to 21CFR820
  • 20 L and 200 L ready-to-use single-use bioprocessing containers

Flexibility

Gibco NaOH solutions provide outsourcing efficiencies for customers manufacturing internally but seeking to partially outsource, as well as for customers with batch sizes that do not justify custom ordering and customers already outsourcing to Thermo Fisher Scientific but seeking additional flexibility. They are produced in the most commonly used concentrations and single-use containers.

Applications of Gibco NaOH solutions

  • pH adjustment
  • Facility/equipment sanitization
  • Chromatography column cleaning, sanitization and storage
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point100°C
CAS1310-73-2
Chemical Name or MaterialSodium Hydroxide
ColorClear
Formula Weight40
Melting Point0°C
Molecular FormulaNaOH
PackagingBioprocess Container
Shelf Life12 months
SynonymCaustic soda, Acidity regulatory, Lye or Soda lye
Concentration0.1 M
GradeCompendial
Percent Purity95 to 100%
Quantity200 L
pH≥12 to ≤14
Unit SizeEach