Sodium Hydroxide (NaOH) Solutions, 0.1–1.0 M
Sodium Hydroxide (NaOH) Solutions, 0.1–1.0 M
Sodium Hydroxide (NaOH) Solutions, 0.1–1.0 M
Gibco™

Sodium Hydroxide (NaOH) Solutions, 0.1–1.0 M

Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantityConcentration
A478290120 L1.0 M
A478260120 L0.1 M
A4782602200 L0.1 M
A478280120 L0.5 M
A4782802200 L0.5 M
A4782902200 L1.0 M
Número do catálogo A4782901
Preço (BRL)
-
Quantity:
20 L
Concentration:
1.0 M

Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing. Available in three different grades and two off-the-shelf sizes, these solutions provide outsourcing efficiencies to help reclaim time, space, and other core resources.

Features of Gibco NaOH solutions include:

  • 0.1 M, 0.5 M, and 1 M concentrations
  • Multi-compendial grade raw materials
  • cGMP manufacturing to 21CFR820
  • 20 L and 200 L ready-to-use single-use bioprocessing containers

Flexibility

Gibco NaOH solutions provide outsourcing efficiencies for customers manufacturing internally but seeking to partially outsource, as well as for customers with batch sizes that do not justify custom ordering and customers already outsourcing to Thermo Fisher Scientific but seeking additional flexibility. They are produced in the most commonly used concentrations and single-use containers.

Applications of Gibco NaOH solutions

  • pH adjustment
  • Facility/equipment sanitization
  • Chromatography column cleaning, sanitization and storage
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificações
Boiling Point100°C
CAS1310-73-2
Chemical Name or MaterialSodium Hydroxide
ColorClear
IUPAC Namesodium hydroxide
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
Melting Point0°C
Molecular FormulaHNaO
PackagingBioprocess Container
SMILES[OH-].[Na+]
Shelf Life12 months
SynonymCaustic soda, Acidity regulatory, Lye or Soda lye
Concentration1.0 M
GradeCompendial
Molecular Weight (g/mol)40.00
Percent Purity95 to 100%
Quantity20 L
pH≥12 to ≤14
Unit SizeEach