Pierce™ Yeast Digest Standard
Thermo Scientific™
Pierce™ Yeast Digest Standard
The Thermo Scientific™ Pierce Yeast Digest Standard is a lyophilized yeast peptide mixture ideal for monitoring LC/MS system performance.
Have Questions?
Catalog NumberQuantity
A4795120 μg
Catalog number A47951
Price (USD)
180.00
Each
Quantity:
20 μg
Price (USD)
180.00
Each
The Thermo Scientific™ Pierce Yeast Digest Standard is a lyophilized yeast peptide mixture ideal for monitoring LC/MS system performance.
Pierce Yeast Digest Standard features include:
  • Positive control sample – well-characterized eukaryotic proteome sample protein digest (>3,000 proteins) for LC/MS method standardization, development, and troubleshooting
  • Verified peptide quality – digestion conditions optimized for minimal missed cleavages or peptide modifications during processing
  • Rigorously tested – high-quality, consistent protein digest guaranteed and documented via lot-specific certificates of analysis
  • Convenient – ready-to-use lyophilized format
The Pierce Yeast Digest Standard is specifically formulated for LC/MS experiments and does not contain salts or detergents. The protein digest is derived from Saccharomyces cerevisiae yeast strain BY4741 (genetic background: MATa his3delta1 leu2delta0 met15delta0 ura3delta0). The yeast proteome is well-documented and characterized, and the digest is an ideal standard for more complex proteome mass spectrometry applications or when a non-mammalian digeste is required for analysis.
Using the standard routinely before analysis of complex samples makes it possible to monitor and normalize LC/MS performance between samples and over time.
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Chemical Name or MaterialYeast peptide digest
ColorWhite
PackagingGlass vial
Physical FormLyophilized
Quantity20 μg
SynonymYeast peptide digest
GradeMS
Unit SizeEach
Frequently asked questions (FAQs)
Which LC/MS analysis process does Yeast Digest Standard help monitor?
Yeast Digest Standard is suitable for monitoring the Ion Source parameters (spray stability, source temperature of ionization, in-source fragmentation) and Mass Analyzer components (mass accuracy from scan to scan) of the LC/MS analysis.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.