Pierce™ HeLa Digest/PRTC Standard
Pierce™ HeLa Digest/PRTC Standard
Pierce™ HeLa Digest/PRTC Standard
Thermo Scientific™

Pierce™ HeLa Digest/PRTC Standard

The Thermo Scientific™ Pierce HeLa Digest/PRTC Standard is a validated complex mammalian protein digest containing additional PRTC heavy isotope-labeled peptides designed for LC/MS performance testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A479975 x 10 μg
A4799610 μg
Catalog number A47997
Price (USD)
726.65
Online exclusive
816.00
Save 89.35
Each
Quantity:
5 x 10 μg
Price (USD)
726.65
Online exclusive
816.00
Save 89.35
Each

The Thermo Scientific™ Pierce HeLa Digest/PRTC Standard is a validated complex mammalian protein digest containing additional PRTC heavy isotope-labeled peptides designed for LC/MS performance testing.

Features of HeLa Digest/PRTC Standard include:

  • Convenient – pre-mixed formulation of HeLa digest and PRTC standard in a ready-to-use format
  • Informative – enables robust performance assessment of LC/MS instruments
  • Verified – high-quality, consistent mixture documented via lot-specific certificates of analysis
  • Stable – provided in a lyophilized format

The Pierce HeLa Digest/PRTC Standard is a lyophilized mixture of 10 μg of HeLa protein digest with 5 pmol PRTC peptides to assess chromatography MS instrument performance. The protein digest is derived from a well-established adenocarcinoma reference HeLa S3 cell line that expresses over 15,000 proteins with relevant post-translational modifications. This complex digest is an ideal standard for complex proteome mass spectrometry applications.

The included Pierce Peptide Retention Time Calibration Mixture contains 15 synthetic heavy isotope-labeled peptides mixed at an equimolar ratio that elute across the chromatographic gradient. The observed retention times and hydrophobicity factors (HFs) for these calibrant peptides are fit to a linear equation to determine the slope of the retention time/HF relationship. This equation and the HF of an uncharacterized peptide are then used to predict retention time. In combination, the mixture is a powerful tool to optimize and assess LC parameters, identify the total peptide elution window, and optimize and monitor MS instrumentation.

This standard is specifically formulated for LC/MS experiments and does not contain salts or detergents. Using the standard routinely before analysis of complex samples makes it possible to monitor and normalize LC/MS performance between samples and over time.

Applications:

  • Qualitative LC assessment
  • QC assay tool for assessment of LC/MS system performance
  • LC/MS standardization
  • LC/MS method development and optimization
  • Normalize results for variation in retention times and peak intensities between runs
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Chemical Name or MaterialHeLa Digest/PRTC Standard
PackagingLyophilized in glass vial
Quantity5 x 10 μg
SynonymHeLa/PRTC
GradeMS
Unit SizeEach

Frequently asked questions (FAQs)

What is the composition of HeLa Digest/PRTC Standard?

HeLa Digest/PRTC Standard is a combination of Pierce HeLa Protein Digest Standard (Cat. No. 88328 and 88329) and Pierce Peptide Retention Time Calibration Mixture (PRTC) (Cat. No. 88321). HeLa Digest/PRTC Standard contains 10 µg of HeLa Digest Standard and 5 pmol of PRTC.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

Which LC/MS analysis process does HeLa Digest/PRTC Standard help monitor?

HeLa Digest/PRTC Standard helps monitor all the steps of LC/MS analysis: LC, Ion Source, and Mass Analyzer. HeLa Digest monitors Ion Source and Mass Analyzer, while PRTC monitors the LC component of the LC/MS analysis process.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.