MagMAX™ Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit - FAQs
MagMAX™ Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit - FAQs
View additional product information for MagMAX™ Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit - FAQs (A48383R)
1 product FAQs found
Do you offer the Proteinase K that is included in the MagMAX Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit (Cat. No. A48383) as a stand-alone product?
No standalone components are validated for In Vitro Diagnostic (IVD) use with this kit.