What alternative do you offer for KingFisher Deep Well 96 Tip Comb, 100 PCS (Compatible with Flex and Presto) (Cat. No. A48438)?
The alternative is KingFisher Plastics for 96 deep‐well format (Cat. No. 97002534). This product is a 96 tip comb for deep‐well magnets that can be used on the KingFisher Flex and Presto instruments.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.