AccuSEQ™ Real-Time PCR Detection Software v3.1
Product Image
Applied Biosystems™
AccuSEQ™ Real-Time PCR Detection Software v3.1
AccuSEQ Real-Time PCR Detection Software is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing usingMás información
Have Questions?
Número de catálogoCantidad
A485091 Kit
Número de catálogo A48509
Precio (CLP)
-
Cantidad:
1 Kit
AccuSEQ Real-Time PCR Detection Software is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing using the SEQ family of real-time PCR assays, including MycoSEQ, resDNASEQ, and ProteinSEQ assays. The software is designed to work with the Applied Biosystems QuantStudio 5 Real-Time PCR Instrument with 0.1-mL, 96-well plates for either Fast or Standard cycling. This particular software is provided via a link for online download.
Features of AccuSEQ Real-Time PCR Detection Software include:
• Automated presence/absence calls for Mycoplasma detection
• Accurate quantitation of host cell line residual DNA
• Non-linear curve fitting of ProteinSEQ assay data for Protein A
• Single software platform for all current and future SEQ real-time PCR assays
• Security, audit, and e-signature capabilities to help enable 21 CFR Pt 11 compliance
Mycoplasma detection
For rapid and accurate Mycoplasma detection, AccuSEQ software offers a plug-in analysis module for automated presence/absence calls when using the MycoSEQ Mycoplasma Detection system. Automated analysis tools enable one-click processing of MycoSEQ assay data, helping deliver presence or absence calls within seconds of data collection being completed.
Residual DNA quantitation
For host cell residual DNA quantitation, a custom module with next-generation algorithms can be used with the resDNASEQ CHO Residual DNA Assay for accurate quantitation.
Protein quantitation
AccuSEQ software helps enable rapid and easy analysis of ProteinSEQ protein quantitation system data without the hassle of multiple exporting steps and reliance on third party software.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (aplicación)PCR en tiempo real (PCRq)
Para utilizar con (equipo)QuantStudio™ 5, 0.1 mL Block Instrument
Línea de productosAccuSEQ
Cantidad1 Kit
Condiciones de envíoTemperatura ambiente
Tipo de softwareReal-Time PCR Software
Tipo de productoReal Time PCR Detection Software
Unit SizeEach
Contenido y almacenamiento
Product letter and License registration code kit
Preguntas frecuentes
When using the AccuSEQ Real-Time PCR Detection Software v3.1 (Cat. No. A48509), for the double signature function, do the users who will sign have to have different roles or could they also hold the same role?
With the AccuSEQ Real-Time PCR Detection Software v3.1 (Cat. No. A48509), the administrator can define whether 1 or 2 signatures are required to sign an experiment. However, it is not possible to specify that the signatures have to come from different roles. The administrator can only define how many signatures (up to 10) are required to sign an experiment.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.