VeritiPro™ Thermal Cycler, 96-well
Applied Biosystems™

VeritiPro™ Thermal Cycler, 96-well

The Applied Biosystems VeritiPro Thermal Cycler delivers proven reliability with advanced temperature control technology and connectivity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeWarrantyFormat
A48769Thermal Cycler with Plastic Package3 Year Extended Warranty, REX warrantyEnduraPlates
A48141Thermal Cycler2 Years96-well
A47397Thermal Cycler with Plastic Package2 YearEnduraPlates
A48768Thermal Cycler with Plastic Package3 Year Extended Warranty, ABRC warrantyEnduraPlates
A47398Thermal Cycler2 YearTriFlex Plates
A48770Thermal Cycler with Plastic Package3 Year Extended Warranty, ABRC warrantyTriFlex Plates
A48771Thermal Cycler with Plastic Package3 Year Extended Warranty, REX warrantyTriFlex Plates
A47396Thermal Cycler with Plastic Package2 Year8-tube Strips
A48766Thermal Cycler with Plastic Package3 Year Extended Warranty, ABRC warranty8-tube Strips
A48767Thermal Cycler with Plastic Package3 Year Extended Warranty, REX warranty8-tube Strips
Catalog number A48769
Price (USD)
12,408.00
Each
Product Type:
Thermal Cycler with Plastic Package
Warranty:
3 Year Extended Warranty, REX warranty
Format:
EnduraPlates
Price (USD)
12,408.00
Each

The Applied Biosystems VeritiPro Thermal Cycler delivers proven reliability with advanced temperature control technology and connectivity. Take advantage of next-level PCR optimization with the precision offered by VeriFlex block technology. Connect to the cloud-enabled VeritiPro Thermal Cycler remotely. Conveniently design and securely upload your methods, monitor runs, and check instrument availability from any mobile device or desktop computer using Connect, our cloud-based platform.

This system may be purchased with a variety of plastics packages, warranties, or both to best fit laboratory needs.

Temperature optimization through VeriFlex technology

VeriFlex blocks are controlled by multiple Peltier elements and are used to optimize PCR temperature with precision. The VeritiPro Thermal Cycler allows for six zones of independent temperature control, giving you the ultimate control of your PCR workflows.

Remote capabilities

By connecting your VeritiPro Thermal Cycler to Connect, you will have complete control of your instrument while away from the lab. With Connect you can check instrument availability through the mobile app, download experimental report files, receive alerts, and more. Download our Instrument Connect App, updated with this functionality and visit our cloud-based platform, to design and securely upload your methods from any mobile device or desktop computer.

Simulation mode

The VeritiPro Thermal Cycler is equipped with simulation modes, allowing it to be run using the same ramp rates as previous or different instruments. Through this feature you will be able to mimic the ramp rates of the Veriti, Applied Biosystems 2720, Bio-Rad T100, Bio-Rad MyCycler, Bioer XP Cycler, Eppendorf Mastercycler, MJ Research PTC-200, and Takara Dice PCR thermal cyclers.

Our quietest instrument yet

With all the equipment running in the lab, why should your thermal cycler add to the noise level? We have revised the internal instrument technology to provide your lab with a thermal cycler that runs at a noise level of less than 48 dB(A).

8-inch touch-screen control

The improved, intuitive, and responsive touch-screen interface allows you to program with ease and limits the amount of time spent training new users.

Compact footprint

With the increasing popularity of our smaller instruments, we have updated our trusted Veriti Thermal Cycler with a smaller footprint. The VeritiPro Thermal Cycler can fit on just about any workbench with its dimensions of 24.5 cm wide and 46.7 cm deep.

Plastics

Applied Biosystems PCR plastics

Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment.

MicroAmp 8-Tube Strips with Attached Domed Caps

MicroAmp 8-Tube Strips with Attached Domed Caps feature individually attached domed caps, dual large end-tabs, and graduated 20-μL markers on each tube to help prevent cross-contamination, pipetting errors, and sample identification errors.

  • Strip of 8 tubes with attached domed caps that open and close independently to prevent contamination and evaporation
  • Angled caps that provide efficient tube strip positioning in racks and thermal cycler blocks
  • Etched A-H letter labeling on individual tubes that is visible when caps are open or closed
  • Dual end tabs for proper labeling and easy handling
  • Graduated 20-μL measuring markers on each tube for visual volume verification

MicroAmp EnduraPlates

MicroAmp EnduraPlates offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. They are a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments. They are validated for use with Applied Biosystems PCR thermal cyclers for optimal fit and performance.

  • Optimized for performance with Applied Biosystems instruments
  • Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
  • Constructed to ANSI/SBS standard, well-suited to high-throughput laboratories and robot handling
  • Available in multiple colors for easy organization of laboratory assays
  • Excellent mechanical stability and flatness for your complete workflow needs

MicroAmp Clear Adhesive Films

MicroAmp Clear Adhesive Films create a secure seal across an entire microplate to prevent evaporation and well-to-well contamination.

  • Adhesive provides tight seal over wells to prevent leaks and evaporation
  • Easy to apply and remove
  • Available as Clear (economical and sticky) or Optical (ready for real-time PCR and non-tacky)

MicroAmp TriFlex 3 x 32-Well PCR Reaction Plates

MicroAmp TriFlex 3 x 32-Well PCR Reaction Plates have a unique design, with three identical polypropylene 32-well PCR plates connected by dual side-tabs for use with 32-well and standard 96-well thermal blocks. The plates offer the same thin-wall construction as all other MicroAmp plastics, for excellent heat transfer from thermal block to sample, and they are validated for use with Applied Biosystems PCR thermal cyclers for optimal fit and performance.

  • Designed for use with the ProFlex 3 x 32-well PCR System and all Applied Biosystems standard 96-well thermal cyclers
  • Easy-to-tear dual side-tabs for simple separation into three individual 32-well plates
  • Semi-skirted for easy handling and labeling
  • Standard 250 μL working volume, with a 300 μL maximum well volume when used with an adhesive film
  • Alphanumeric labeling on each 32-well plate for easy well identification
  • Certified human DNA-, DNase-, and RNase-free

MicroAmp Clear Adhesive Films

MicroAmp Clear Adhesive Films create a secure seal across an entire microplate to prevent evaporation and well-to-well contamination.

Warranties

Rapid Exchange (REX) extended warranty

In most countries, your thermal cycler comes with a standard two-year warranty. The package offered here provides additional warranty coverage to your existing manufacturer's limited warranty. With the Rapid Exchange (REX) extended warranty, if your instrument needs repair, just call our Support Center and we will exchange it for a factory-certified refurbished replacement instrument within one business day from the time we determine a replacement is required. Instruments are typically received within three days of shipment and are yours to keep. All parts, labor, and shipping charges are included, and existing warranty contracts will be transferred to replacement units.

The Rapid Exchange extended warranty is ideal for customers with user-installable instruments that do not require asset tag consistency, and for those operating under critical timelines.

AB Repair Center (ABRC) extended warranty

In most countries, your thermal cycler comes with a standard two-year warranty. The package offered here provides additional warranty coverage to your existing manufacturer's limited warranty. With ABRC support, factory-trained engineers will perform maintenance or repair your instrument at our remote service center. Just send us your instrument and we will maintain or repair it and ship it back to you typically within three weeks. All parts, labor, and shipping charges are included.

The ABRC extended warranty is ideal for customers in regulated environments who must maintain asset tag consistency to comply with regulated protocols.

Certain exclusions apply. Contact your Service and Support Representative for details.

Specifications
DescriptionVeritiPro™ Thermal Cycler + Extended Warranty + Plastics Package, 96-well, REX warranty, EnduraPlates
Dimensions18.3 x 9.6 x 8.5 in. (46.5 x 24.5 x 21.7 cm)
Display Type8 in. Color TFT LCD Touchscreen
For Use With (Equipment)Thermal Cycler
FormatEnduraPlates
Frequency50 to 60 Hz
Max. Ramp Rate6°C/sec.
Memory16 Gb Onboard Memory
Quantity1 Package
Thermal Accuracy±0.25°C (35°C to 99.9°C)
Thermal Range0°C to 100°C
Thermal Uniformity<0.5°C (30 sec. after reaching 95°C)
User InterfaceUSB, Wi-Fi, Ethernet
Voltage100/240 V
Warranty3 Year Extended Warranty, REX warranty
Wattage600 W
Weight26.5 lb. (12 kg)
Block Configurations96-well, 6-Zone VeriFlex Block
Capacity96-well
No. of Programs>1000
Product TypeThermal Cycler with Plastic Package
Unit SizeEach
Package Components
A47397
Catalog Number
A47397
Unit Size
Qty
1
ZGEXSCVERITIP96
Catalog Number
ZGEXSCVERITIP96
Unit Size
Qty
1
Contents & Storage
• VeritiPro Thermal Cycler + Extended Warranty + Plastics Package
• Country-specific power cord
• 1 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plate with Barcode
• 1 x MicroAmp Clear Adhesive Film
• 1 x MicroAmp 32-well Clear Adhesive Film
• Rapid Exchange (REX) Support Plan

Frequently asked questions (FAQs)

Have you tested the Veriti and VeritiPro Thermal Cyclers in low humidity conditions (below 20%)?

The recommended conditions are determined to ensure no static discharge (low humidity) occurs.
Recommended Conditions:
- Veriti: 20-80% humidity
- VeritiPro: 15-80% humidity
We have not tested Veriti and VeritiPro thermal cyclers in low humidity, below 20%, therefore we cannot recommend using them in those conditions.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

I get the following error message on my Veriti Pro: ‘The instrument is due for servicing for the following test(s): Temperature Non-Uniformity test, Temperature Verification Test, Cycle performance Test, Heated Cover Test’. Do I need to do these tests?

The error message on the Thermal Cycler is a reminder that regular maintenance is useful to ensure the optimal functioning of the appliance. You can switch off these reminders or change the interval in the device settings. On the VeritiPro you will find the menu item “Service Reminders” in “Maintenance and Service” section of the user guide (page 58).

Find additional tips, troubleshooting help, and resources within our PCR Thermal Cyclers Support Center.