MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0, Lite - FAQs
MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0, Lite - FAQs
View additional product information for MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0, Lite - FAQs (A49380)
3 product FAQs found
What are the recommended system requirements for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0, Lite?
Here are the recommended system requirements for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0, Lite:
- Intel Core i5 Processor, 2.4 GHz or faster
- Microsoft Windows 10 OS 64-bit, 16 GB RAM, at least 250 GB for software installation and data storage
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the difference between MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 vs MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0, Lite?
Use the Lite version if you plan to perform simple analysis for use on a stand-alone PC (not connected to the SeqStudio Genetic Analyzer). Use the System version if you plan to run complete analysis on the SeqStudio Genetic Analyzer.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the catalog number for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0, Lite?
The catalog number for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0, Lite is Cat. No. A49380.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.