MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0 - FAQs
MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0 - FAQs
View additional product information for MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0 - FAQs (A49382)
7 product FAQs found
What are the recommended system requirements for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 (System version)?
Here are the recommended system requirements for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 (System version):
- Intel Core i7 Processor, 3.1 GHz or faster
- Microsoft Windows 10 OS, 64-bit, 16 GB RAM, and at least 250 GB for software installation and data storage
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the key features of MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0?
- Compatibility with SeqStudio Genetic Analyzer
- Support for SeqStudio Genetic Analyzer Protocol: AB_KB_3200_SeqStudio_POP1_BDTv1
- New KB Basecaller V1.4.2.5
- Windows 10, 64-bit compatibility
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What libraries are compatible with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0?
- MSID 16S rDNAFull Gene Library v2.0
- MSID Fungal library v2018
- MicroSEQ_ID_16S_rdna_500_Library_v2019
- MicroSEQ_ID_16S_rdna_500_Supplemental_Library_v2019
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the specifications of the computer that is provided with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0?
Here are the specifications of the computer that is provided with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0:
Computer: Intel Core i7-8700 CPU@3.20 Ghz, with 16 GB of RAM and a minimum of 4 GB hard disk space
Operating System: Windows 10; 64-bit processor
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Does a computer come with the MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0?
The MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 is shipped with a computer.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the difference between MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 vs MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0, Lite?
Use the Lite version if you plan to perform simple analysis for use on a stand-alone PC (not connected to the SeqStudio Genetic Analyzer). Use the System version if you plan to run complete analysis on the SeqStudio Genetic Analyzer.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the catalog number for MicroSEQ ID Software (digital download) for SeqStudio Genetic Analyzer v1.0?
The catalog number for MicroSEQ ID Software (digital download) for SeqStudio Genetic Analyzer v1.0 is A49382.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.