Tuberculin PPD Kit
Tuberculin PPD Kit
Tuberculin PPD Kit
Tuberculin PPD Kit
Applied Biosystems™

Tuberculin PPD Kit

The standard method for detection of Tuberculosis is the tuberculin test, where a small amount of antigen is injected into the skin and the immune reaction (swelling) is measured. The Tuberculin PPD Kit has both Bovine & Avian Tuberculin PPD.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySufficient For
A4942240 x 2 mL Vials800 Doses (400 doses of Avian PPD, 400 doses of Bovine PPD)
A4942520 x 5 mL Vials1000 Doses (500 doses of Avian PPD, 500 doses of Bovine PPD)
Catalog number A49422
Price (USD)
-
Quantity:
40 x 2 mL Vials
Sufficient For:
800 Doses (400 doses of Avian PPD, 400 doses of Bovine PPD)

Bovine tuberculosis (bTB) is a notifiable disease caused by the bacteria Mycobacterium bovis which infects the lungs and associated lymph nodes although lesions can be found in any organ or body cavity of diseased animals. Infection is often sub-clinical. bTB has a prolonged course and symptoms take months-years to appear making the diagnosis of the disease complex.

It is found primarily in cattle but can also be found in cervids (deer and elk), bison, goats, carnivores (coyotes) and omnivores (badgers); any warm-blooded animal including humans can be affected by the disease. Transmission of M. bovis within and between species is mainly by the airborne route or by ingestion of the bacterium (in humans by drinking raw milk).

The standard method for detection of Tuberculosis is the tuberculin test, where a small amount of antigen is injected into the skin and the immune reaction (swelling) is measured. The Tuberculin PPD Kit has both Bovine & Avian Tuberculin PPD.

Regulatory requirements vary by country; products may not be available in your geographic area.

Specifications
AssayTuberculin PPD
Concentration3000 IU (Bovine Tuberculin PPD), 2500 IU (Avian Tuberculin PPD)
For Use With (Application)Detection of bovine Tuberculosis
For Use With (Equipment)Syringe/Needle
FormatKit
MaterialPurified Protein Derivative (PPD)
Product TypeTuberculin PPD Kit
Protein FamilyMycobacterium bovis, Mycobacterium avium
Quantity40 x 2 mL Vials
Regulatory StatusVMP (Veterinary Medicinal Product)
Shipping ConditionShipped at 2°C to 37°C
Storage Requirements2°C to 8°C.
SubstratePurified Protein Derivative (PPD)
Sufficient For800 Doses (400 doses of Avian PPD, 400 doses of Bovine PPD)
Target Species ValidatedBovine
Detection MethodImmune Reaction
Immunoassay Kit Format2 Component Kit
Target SpeciesCattle
Unit Size40 x 2 mL vials (800 doses)
Contents & Storage
Store at 2°C to 8°C.