CTS™ OpTmizer™ Pro SFM, no phenol red
Gibco™
CTS™ OpTmizer™ Pro SFM, no phenol red
Gibco™ CTS OpTmizer Pro SFM is a novel serum-free medium (SFM) developed for the growth and expansion of human T lymphocytes (e.g., CD4+, CD8+, polyclonal, or antigen-specific) used in allogeneic cell therapies.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
A49661055 L
A496610710 L
Número de catálogo A4966105
Precio (USD)
-
Cantidad:
5 L
Gibco™ CTS OpTmizer Pro SFM is a novel serum-free medium (SFM) developed for the growth and expansion of human T lymphocytes (e.g., CD4+, CD8+, polyclonal, or antigen-specific) used in allogeneic cell therapies. This medium improves central memory phenotype and cell growth by shifting the cell's metabolism. The result is a larger population of central memory cells in a shorter period of time.
CTS OpTmizer Pro SFM consists of CTS OpTmizer Pro Basal Medium and CTS OpTmizer Supplement, which are mixed together prior to use.
CTS OpTmizer Pro SFM in 5 L and 10 L bag format is compatible with closed-system processing for media preparation and integration to standard cell therapy equipment, including CTS devices (Xenon, Rotea, and DynaCellect).
Benefits of CTS OpTmizer Pro SFM include:
  • Supplement component now in bag format to allow for closed mixing with basal medium
  • Reduces the number of open steps in your process, thus reducing contamination risks
  • Easy integration with cell therapy equipment, including CTS Xenon, Rotea, and DynaCellect
  • Maintains central memory T cells further into expansion and is well suited for longer workflows (>15 days)
  • Offers >90% (average) T-cell fold-expansion (from healthy donors) compared to CTS OpTmizer medium
  • Modest increases in central memory cells at early stage in expansion protocol result in larger cell yields at harvest
  • Helps achieve the same number of cells in a shorter duration
  • Ideally suited to the production of allogeneic therapies
Features include:
  • Flexible pack sizes in Aegis5-14 film: basal medium in 5 L and 10 L bags
  • Weldable C-Flex and PVC lines
  • Supports allogeneic T-cell processes through all stages of development and manufacturing
  • Supports T-cell activation using Dynabeads magnetic beads, soluble antibodies, and stimulatory antibody-presenting cell protocols
Increased efficiency and consistence performance
CTS OpTmizer Pro SFM, no phenol red, is serum-free and xeno-free and supports high density cultures in static cultures. It displays consistent performance in the support of T-cell expansion with better central memory phenotype in a shorter time.
Enhanced expansion
CTS OpTmizer Pro SFM, no phenol red, increases the number of central memory cells at an early stage in the expansion process and maintains T-cell functionality.
Enhanced design and compatibility
The supplement component is now in a bag with a weldable PVC line, allowing for closed-system mixing with the CTS OpTmizer Pro basal medium bag. This feature, along with larger 5 L and 10 L basal medium sizes, addresses manufacturers' need for scale, efficiency, and contamination risk mitigation.
Use of Thermo Scientific™ Aegis5-14 Film allows compatibility with closed systems. This film is a five-layer cast film produced in a cGMP facility. The outer layer is a polyester elastomer coextruded with an ethyl vinyl alcohol (EVOH) barrier layer and an ultra-low density polyethylene product contact layer. Aegis5-14 Film is manufactured using no animal derived components.
Simplify your transition into clinical research
The Gibco™ Cell Therapy Systems (CTS) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices.
CTS products deliver:
  • Reduced burden in qualifying reagents
  • Appropriate certificates of analysis and origin available
  • Convenient access to our Drug Master File (DMF)
Especificaciones
Tipo de célulaT Cells
Entorno de cultivoBags, Bioreactors
Línea de productosOpTmizer™ Pro
Cantidad5 L
Duración de almacenamiento12 Months
Condiciones de envíoTemperatura ambiente
ClasificaciónXeno-free
Culture TypeStatic Culture
FormularioLíquido
Serum LevelSerum-free
EsterilidadEstéril con filtro
Sterilization Method0.2 μm Sterile-filtered
Con aditivosNone
Sin aditivosNo Phenol Red
Unit SizeEach
Contenido y almacenamiento
• 1 x 5000 mL bag CTS OpTmizer Pro Basal Medium, store at 2°C to 8°C, protect from light
• 1 x 130 mL CTS OpTmizer Expansion Supplement in 250 mL bag, store at 2°C to 8°C, protect from light
Preguntas frecuentes
Are the 5L and 10L sizes/configurations of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107) compatible with CTS devices?
Yes. The 5L and 10L BPCs (bioprocess containers) are weldable to Xenon, Rotea, and DynaCellect consumables.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
With the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107), how much supplement should be added to the basal medium?
The 5L and 10L kits are designed so that the entire volume of the supplement BPC (bioprocess container) is added to the basal medium BPC.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
With the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107), how is the supplement added to the basal medium?
The supplement BPC (bioprocess container) has 24 in. of sterile weldable 3/32 in. ID, 5/32 in. OD DHEP-free PVC tubing. With fill volumes of 130 mL and 260 mL for the 5L and 10L kits respectively, the supplement BPC can be directly welded to the basal medium BPC.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the differences between the BPCs (bioprocess containers) in the 1L sizes of CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM vs. the 5L and 10L sizes?
In the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM, both the basal medium and supplement components are in BPCs (bioprocess containers) composed of Aegis 5-14 film. The basal medium BPC has PVC and C-Flex lines whereas the supplement BPC has a PVC line.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the stability of the CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM in the new 5L and 10L sizes?
The 5L and 10L sizes of both CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107) have 12-month expiry.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.