CTS™ OpTmizer™ Pro SFM, no phenol red, 10 L - FAQs
CTS™ OpTmizer™ Pro SFM, no phenol red, 10 L - FAQs
View additional product information for CTS™ OpTmizer™ Pro SFM, no phenol red - FAQs (A4966107, A4966105)
5 product FAQs found
Are the 5L and 10L sizes/configurations of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107) compatible with CTS devices?
Yes. The 5L and 10L BPCs (bioprocess containers) are weldable to Xenon, Rotea, and DynaCellect consumables.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
With the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107), how much supplement should be added to the basal medium?
The 5L and 10L kits are designed so that the entire volume of the supplement BPC (bioprocess container) is added to the basal medium BPC.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
With the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107), how is the supplement added to the basal medium?
The supplement BPC (bioprocess container) has 24 in. of sterile weldable 3/32 in. ID, 5/32 in. OD DHEP-free PVC tubing. With fill volumes of 130 mL and 260 mL for the 5L and 10L kits respectively, the supplement BPC can be directly welded to the basal medium BPC.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the differences between the BPCs (bioprocess containers) in the 1L sizes of CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM vs. the 5L and 10L sizes?
In the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM, both the basal medium and supplement components are in BPCs (bioprocess containers) composed of Aegis 5-14 film. The basal medium BPC has PVC and C-Flex lines whereas the supplement BPC has a PVC line.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the stability of the CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM in the new 5L and 10L sizes?
The 5L and 10L sizes of both CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107) have 12-month expiry.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.