What cell counter do you offer that can count live and dead cells, measure cell viability, cell size, has high-resolution touch screen, and comes with reusable slides?
We suggest the Countess 3 and 3 FL Automated Cell Counters with starter packages; please see the product pages linked below:
Countess 3 FL Automated Cell Counter Starter Package + ABRC Extended Warranty (Cat. No. A50303)
Countess 3 FL Automated Cell Counter Starter Package + REX Extended Warranty (Cat. No. A50299)
Countess 3 FL Automated Cell Counter Starter Package (Cat. No. A49866)
Countess 3 Automated Cell Counter Starter Package + ABRC Extended Warranty (Cat. No. A50302)
Countess 3 Automated Cell Counter Starter Package + REX Extended Warranty (Cat. No. A50298)
Countess 3 Automated Cell Counter Starter Package (Cat. No A49865)
For more information about out Countess 3 Automated Cell Counters See the brochure linked below:
linkCountess 3 Automated Cell Counters: A counter that thinks like a cell biologist
Countess 3 FL Automated Cell Counter Starter Package + ABRC Extended Warranty (Cat. No. A50303)
Countess 3 FL Automated Cell Counter Starter Package + REX Extended Warranty (Cat. No. A50299)
Countess 3 FL Automated Cell Counter Starter Package (Cat. No. A49866)
Countess 3 Automated Cell Counter Starter Package + ABRC Extended Warranty (Cat. No. A50302)
Countess 3 Automated Cell Counter Starter Package + REX Extended Warranty (Cat. No. A50298)
Countess 3 Automated Cell Counter Starter Package (Cat. No A49865)
For more information about out Countess 3 Automated Cell Counters See the brochure linked below:
linkCountess 3 Automated Cell Counters: A counter that thinks like a cell biologist