SeqStudio™ Genetic Analyzer QST with SAE - FAQs
SeqStudio™ Genetic Analyzer QST with SAE - FAQs
View additional product information for SeqStudio™ Genetic Analyzer QST with SAE - FAQs (A49988)
1 product FAQs found
What is the catalog number for SeqStudio Genetic Analyzer QST with SAE?
The catalog number for SeqStudio Genetic Analyzer QST with SAE is A49988.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.