CTS™ Xenon™ MultiShot Electroporation Cartridge
Gibco™

CTS™ Xenon™ MultiShot Electroporation Cartridge

The CTS Xenon MultiShot Electroporation Cartridge is for use with the Xenon Electroporation System. The MultiShot cartridge has sample processing capacity of 5—25 mL.
Have Questions?
Catalog NumberQuantity
A503061 Cartridge
Catalog number A50306
Price (MXN)
-
Quantity:
1 Cartridge

The CTS Xenon MultiShot Electroporation Cartridge is for use with the Xenon Electroporation System. The single-use cartridge has an electroporation chamber made of Tritan™ MX711 copolyester with gold-plated electrodes and is sterilized by gamma irradiation. It can process 1 x 108 to 2.5 x 109 T cells in 5–25 mL of sample volume. The MultiShot cartridge enables electroporation in a closed system with the flexibility of sterile welding to either PVC or CFlex™ tubing. Promising electroporation conditions that have been optimized in the Neon Transfection System can be scaled to the Xenon Electroporation System without sacrificing performance.

  • The Gibco CTS Xenon MultiShot Electroporation Cartridge is for use with the Xenon Electroporation System for cell therapy research and development
  • The single-use cartridge has an electroporation chamber made of Tritan™ MX711 copolyester with gold-plated electrodes and is sterilized by gamma irradiation
  • It can process 1 x 108 to 2.5 x 109 T cells in a closed system with the flexibility of sterile welding to either PVC or CFlex™ tubing
  • Promising electroporation conditions that have been optimized in the Neon Transfection System can be scaled to the Xenon Electroporation System without sacrificing performance
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
DisposableSingle-use
Manufacturing QualityISO 13485
Quantity1 Cartridge
Shelf Life24 Months from Date of Manufacture
Sterilization MethodGamma Irradiation
Capacity (Metric)5 to 25 mL
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
For Use With (Equipment)Gibco CTS Xenon Electroporation System
MaterialTritan MX711 Copolyester
Unit SizeEach