Genexus™ Dx Library Strips 1 and 2-HD
Ion Torrent™
Genexus™ Dx Library Strips 1 and 2-HD
Kit provides eight each of the strips required to perform automated library preparation using the Ion Torrent™ Genexus™ Dx Integrated Sequencer.
Have Questions?
Número de catálogoN.º de reacciones
A5043032 Reactions
Número de catálogo A50430
Precio (MXN)
-
N.º de reacciones:
32 Reactions
The Ion Torrent Genexus Dx Library Strips 1 and 2-HD kit provides eight each of the strips required to perform automated library preparation using the Genexus Dx Integrated Sequencer. This kit, combined with Genexus Dx Barcodes 1-32 HD, supports automated preparation and normalization of up to 32 single-pool Ion AmpliSeq HD libraries.
The Genexus Dx Integrated Sequencer is an in vitro diagnostic (IVD) medical device intended to be used by trained personnel in a professional laboratory environment for automated, qualitative targeted sequencing of human DNA or RNA extracted from formalin-fixed, paraffin-embedded (FFPE) tumor tissue samples or plasma samples.
For In Vitro Diagnostic Use. Not available in all countries or regions.
Especificaciones
Para utilizar con (aplicación)Secuenciación
Para utilizar con (equipo)Genexus™ Dx Integrated Sequencer
BibliotecasTargeted Sequencing Library
N.º de reacciones32 Reactions
N.º de muestras32 Samples
Tipo de ácido nucleicoDNA, RNA, and cfTNA* (*Not available in the U.S.)
Línea de productosGenexus™ Dx
Tipo de productoLibrary Strip
Cantidad1 Ea.
Condiciones de envíoAprobado para su envío en hielo húmedo o seco
Unit SizeEach
Contenido y almacenamiento
• Genexus Strip 2-HD, store at -10 to -30°C
• Genexus Strip 1, store at 2–8°C
Preguntas frecuentes
How should I prep Genexus Dx Library Strips 1 and 2-HD for use on the Genexus Dx platform?
Remove the library strips from their boxes in the refrigerator or freezer, and prepare them for loading in the sequencer.
- Genexus Dx Strip 1 needs to equilibrate to room temperature for 30 mins.
- Genexus Dx Strip 2-HD needs to thaw on ice for at least 30 mins. Keep the strips on ice until you load them in the sequencer. Confirm that the strip contents are completely thawed before installing in the sequencer.
Genexus Dx Strip 1 contains magnetic beads in one or two positions, yellow or brown in color, that sometimes get trapped in the upper "keyhole" of the tube. Dislodge these beads from the keyhole by inverting each strip 3-4 times, before installing the strip in the sequencer.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the storage conditions for Genexus Dx Library Strips 1 and 2-HD (Cat. No. A50430)?
Genexus Dx Strip 1 (red) should be stored at 4 degrees C and Genexus Dx Strip 2-HD (purple) should be stored at -20 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many libraries can I make using the Genexus Dx Library Strips 1 and 2-HD (Cat. No. A50430)?
Genexus Dx Library Strips 1 and 2-HD (Cat. No. A50430) for Ion AmpliSeq HD library-based chemistry provides eight pairs of strips/kit, to perform automated library preparation using the Genexus Dx Integrated Sequencer.
Each library strip is needed to create an Ion Ampliseq HD library and supports up to 4 barcodes.
So 8 strip pairs x 4 = up to 32 DNA or RNA samples/run
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many strips are contained in the Genexus Dx Library Strips 1 and 2-HD (Cat. No. A50430)?
Genexus Dx Library Strips 1 and 2-HD (Cat. No. A50430) for Ion AmpliSeq HD library-based chemistry provides eight pairs of strips/kit, to perform automated library preparation using the Genexus Dx Integrated Sequencer.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.