Genexus™ Dx Sequencing Kit
Ion Torrent™
Genexus™ Dx Sequencing Kit
The Ion Torrent Genexus Dx Sequencing Kit provides the materials required to perform two initializations and automated sequencing of twoMás información
Have Questions?
Número de catálogoCantidad
A504322 procesamientos
Número de catálogo A50432
Precio (MXN)
70,984.84
Each
Cantidad:
2 procesamientos
Precio (MXN)
70,984.84
Each
The Ion Torrent Genexus Dx Sequencing Kit provides the materials required to perform two initializations and automated sequencing of two multilane chips using the Genexus Dx Integrated Sequencer.
The Genexus Dx Integrated Sequencer is an in vitro diagnostic (IVD) medical device intended to be used by trained personnel in a professional laboratory environment for automated, qualitative targeted sequencing of human DNA or RNA extracted from formalin-fixed, paraffin-embedded (FFPE) tumor tissue samples or plasma samples.
For In Vitro Diagnostic Use. Not available in all countries or regions.
Especificaciones
Molde controlGenexus Dx Bottle 2
Tipo de controlGenexus Dx Bottles 1 and 3
Para utilizar con (aplicación)Secuenciación
Para utilizar con (equipo)Genexus™ Dx Integrated Sequencer
Línea de productosGenexus™ Dx
Tipo de productoSequencing Kit
Cantidad2 procesamientos
Condiciones de envíoQualified for Ambient or Wet Ice
Unit SizeEach
Contenido y almacenamiento
• Genexus Dx Cartridge, store at -10 to -30°C
• Genexus Dx Bottle 2, store at room temperature
• Genexus Dx Bottles 1 and 3, store at room temperature
Preguntas frecuentes
How long are the Genexus Dx Sequencing Kit consumables stable on the Genexus Dx instrument?
The Genexus Dx Sequencing Kit consumables are stable on the Genexus Dx instrument for up to two weeks after initialization, allowing flexibility to use the chip lanes according to sample throughput needs.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many initializations can I perform with the Genexus Dx Sequencing Kit?
The Genexus Dx Sequencing Kit provides the materials required to perform two initializations and automated sequencing of two multilane chips using the Genexus Dx Integrated Sequencer.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the contents and storage conditions for the Genexus Dx Sequencing Kit (Cat. No. A50432)?
Here are the contents and storage conditions:
- Genexus Dx Cartridge, store at -10 to -30 degrees C
- Genexus Dx Bottle 2, store at room temperature
- Genexus Dx Bottles 1 and 3, store at room temperature
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.