CTS™ B-27™ Supplement, XenoFree, minus vitamin A (50X)
Gibco™

CTS™ B-27™ Supplement, XenoFree, minus vitamin A (50X)

CTS B-27 Supplement, XenoFree, minus vitamin A, is based on the classic serum-free B-27 supplement and designed to help meet regulatory requirements for cell therapy manufacturing.
Have Questions?
Catalog NumberQuantity
A504760110 mL
Catalog number A5047601
Price (USD)
354.00
Each
Quantity:
10 mL
Price (USD)
354.00
Each

Gibco CTS B-27 Supplement, XenoFree, minus vitamin A, is a cGMP-manufactured, serum-free supplement designed to meet regulatory requirements for cell therapy manufacturing. It supports the growth and differentiation of a variety of cell types derived from pluripotent stem cells (PSCs). The absence of vitamin A makes this formulation ideal for the cultivation and expansion of neural progenitor and stem cells without inducing differentiation until desired.

CTS quality

Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Accompanied by documentation to support your regulatory filing

  • Access to our Drug Master File (DMF) or Regulatory Support File (RSF)
  • Traceability documentation—Certificate of Origin, Certificate of Analysis, and Safety Data Sheet

Reduced risk from animal-origin materials and human-derived components

CTS B-27 Supplement, XenoFree, minus vitamin A, is formulated with human-derived components and AOF recombinant components. To help reduce viral clearance risks, the use of human-derived components has been minimized and human-derived components are limited to materials that help meet regulatory standards for donor qualification, viral testing, and viral inactivation.

CTS B-27 Supplement, XenoFree, minus vitamin A, was specifically designed for use in cell therapy applications by making targeted modifications to the classic serum-free B-27 Supplement. CTS B-27 Supplement, XenoFree, minus vitamin A, supports the robust growth, expansion, and differentiation of stem cells, making it a valuable addition to any cell therapy protocol where cGMP standards are required.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Concentration50X
Culture TypeCells derived from Pluripotent Stem Cells (PSCs)
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity10 mL
Shelf Life12 Months from Date of Manufacture
Shipping ConditionDry Ice
ClassificationSerum-free, Xeno-free
FormLiquid
Product TypeCell Culture Supplement
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
Store at -5°C to -20°C and protect from light.

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

Do you offer CTS media that are compatible with CTS B-27 Supplement Xeno-Free?

Yes, examples include CTS Neurobasal Medium (Cat. No. A13712), CTS Neurobasal-A Medium (Cat. No. A13710), and CTS KnockOut DMEM/F12 (Cat. No. A13708).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How do I request the traceability documentation for CTS B-27 Supplement Xeno-Free?

For instructions on how to request traceability documentation, please visit the Cell Therapy Systems Support webpage (https://www.thermofisher.com/us/en/home/clinical/cell-gene-therapy/cell-therapy/cell-therapy-systems/cell-therapy-systems-support.html).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What accompanying documentation is available for CTS B-27 Supplement Xeno-Free?

Traceability documentation including FDA Drug Master Files and/or Regulatory Support Files, certificates of analysis, and certificates of origin are available upon request.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What varieties of CTS B-27 Supplement Xeno-Free are available?

CTS B-27 Supplement Xeno-Free is available in three versions:
- CTS B-27 Supplement Xeno-Free (Cat. No. A5047501)
- CTS B-27 Supplement Xeno-Free minus Vitamin A (50X) (Cat. No. A5047601)
- CTS B-27 Supplement Xeno-Free minus Insulin (50X) (Cat. No. A5047701)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.