Spectral Calibration Plate with Alexa 647 Dye, 96-well
Thermo Scientific™

Spectral Calibration Plate with Alexa 647 Dye, 96-well

The 96-well Spectral Calibration Plate with Alexa Fluor 647 is used to maintain the QuantStudio Dx, QuantStudio 5 Dx, and QuantStudio 7 Pro Dx real-time PCR instruments with 96-well blocks.
Have Questions?
Catalog NumberQuantity
A505511 Plate
Catalog number A50551
Price (USD)
213.00
Each
Quantity:
1 Plate
Price (USD)
213.00
Each

The 96-well Spectral Calibration Plate with Alexa Fluor 647 is used to maintain the QuantStudio Dx, QuantStudio 5 Dx, and QuantStudio 7 Pro Dx real-time PCR instruments with 96-well blocks. Please see the instrument user guide for full instructions on use.

For In Vitro Diagnostic Use.

Specifications
Calibrated DyeAlexaFluor™ 647
Calibration TypeSpectral Calibration
For Use With (Application)Spectral Calibration
For Use With (Equipment)QuantStudio™ 5 Dx, QuantStudio™ 7 Pro Dx
Format96-well, 0.2 mL
Quantity1 Plate
Shipping ConditionDry Ice
Product TypeCalibration Plate
Unit SizeEach
Contents & Storage
Store at -20°C ± 5°C.