Can you please let me know which plates with and without lid are compatible with the Multiskan SkyHigh Microplate Spectrophotometer?
The Multiskan SkyHigh Microplate Spectrophotometer supports clear bottom 6- to 48-, 96- and 384-well microplates with and without lids, where the maximum height of the microplate or a combination of the microplate and lid is 19.5 mm. Please refer to the Plate Selection Table for Plate Readers brochure (https://assets.thermofisher.com/TFS-Assets/BID/Flyers/nunc-plate-selection-table-plate-readers-flyer.pdf) for specific plates to use.
You can also see the specific plate types in SkanIt Software v7.
You can also see the specific plate types in SkanIt Software v7.