Search
View additional product information for QuantStudio™ 7 Pro Dx Real-time PCR System, 384-well, laptop - FAQs (A51227)
3 product FAQs found
Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.
The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
The issue might be due to the block not being properly seated in the instrument. Please follow the steps below:
It is not possible to add or exchange filters in the QuantStudio 7 Pro Real-Time PCR System.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center