Product Image
Applied Biosystems™
QuantStudio™ 7 Pro Dx Block Upgrade Kit, 96-well, IVD
The IVD QuantStudio 7 Pro Dx Block Upgrade Kit is an optional accessory for the QuantStudio 7 Pro Dx Real-TimeRead more
Have Questions?
Catalog NumberQuantity
A512311 kit
Catalog number A51231
Price (USD)
-
Quantity:
1 kit
The IVD QuantStudio 7 Pro Dx Block Upgrade Kit is an optional accessory for the QuantStudio 7 Pro Dx Real-Time PCR System. Interchangeable blocks enable application versatility with easy, powered-up interchange of block types.
The QuantStudio 7 Pro Dx system accommodates the interchange of 96-well (0.2 mL) and 384-well blocks. The system is delivered with one block type; other block types may be purchased separately.
For In Vitro Diagnostic Use.
Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)QuantStudio™ 7 Pro Dx
Product LineQuantStudio
Quantity1 kit
Shipping ConditionRoom Temperature
Product TypeUpgrade Kit
Unit SizeEach
Contents & Storage
IVD 96-well, 0.2 mL block, IVD 96-well, 0.2 mL heated cover, and accessories