CTS™ Viral Production Medium, 1 L - FAQs
CTS™ Viral Production Medium, 1 L - FAQs
View additional product information for CTS™ Viral Production Medium - FAQs (A5416001, A5416002, A5144002, A5144001, A5144003)
1 product FAQs found
Do I need to supplement CTS Viral Production Medium prior to use?
No, CTS Viral Production Medium already contains 4mM GlutaMAX Supplement and does not need further supplementation.