Viral Production Medium, AGT™, 100 L - FAQs
Viral Production Medium, AGT™, 100 L - FAQs
View additional product information for Viral Production Medium, AGT™ - FAQs (A5147101, A5147102, A5147103, A5147104)
6 product FAQs found
What settings should I use to count my cells using the Vi-CELL XR Cell Counter?
You can refer to page 9 of the AAV-MAX Helper-Free AAV Production System User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0019619_AAV-MAX_Helper-Free_AAV_ProductionSystem_UG.pdf) for detailed Vi-CELL XR settings. We recommend diluting cell count samples with PBS to eliminate precipitate artifacts that may be created by interaction with the Vi-CELL reagents.
Does Viral Production Medium, AGT contain a surfactant? Do I need to add extra surfactant for culturing CTS VPC2.0 in bioreactors ?
While the full formulation for Viral Production Medium, AGT is proprietary information that cannot be disclosed, we can confirm that the medium contains a surfactant and that no additional surfactant would need to be added to the medium to support your suspension CTS VPC2.0 in 2 L bench-top bioreactor to 50 L bioreactor.
What is the concentration of glucose in Viral Production Medium, AGT?
Viral Production Medium, AGT is formulated to contain glucose at a concentration of 6 g/L.
What type of filter is recommended to prepare liquid aseptic media from Viral Production Medium, AGT powder?
Viral Production Medium, AGT powder needs to be reconstituted in deionized distilled water or Water For Injection (WFI) for Cell Culture (Cat. No. A12873) and then immediately sterilized by membrane filtration (positive pressure recommended). A low protein binding filter (such as PVDF) with a pore size of 0.2 µM is recommended.
How does Viral Production Medium, AGT powder differ from the research-grade Viral Production Medium (Cat. No. A48179)?
GlutaMAX Supplement (Cat. No. 35050061) is already present in the one-part Viral Production Medium, AGT powder. Once rehydrated, the formulations of the two products are identical provided the research-grade Viral Production Medium (Cat No. A48179) has been supplemented with GlutaMAX Supplement at point of use.
How much Viral Production Medium, AGT powder do I weigh out for one liter of media?
25.6 grams of Viral Production Medium, AGT powder is required for 1 L of media. We do not advise preparing less than 1 L at a time to ensure consistent weighing and rehydration process.