Why is the processing time for MS analysis less with SureQuant IP-MS Sample Preparation kits compared to traditional IP methods?
SureQuant Protein IP-MS Sample Preparation kits use a trypsin enzyme elution method to dissociate the bound immune complexes from the magnetic beads, which can then be digested directly in-solution and followed up by MS analysis on the same day; no SDS-PAGE purification is required. The entire process take 4 hours compared to traditional IP methods that usually take around 21 hrs.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.