Viral Production Cells 2.0 (CN) - FAQs
Viral Production Cells 2.0 (CN) - FAQs
View additional product information for Viral Production Cells 2.0 (CN) - FAQs (A52021)
5 product FAQs found
Can I use Viral Production Cells (Cat. No. A35347, A35827) with the AAV MAX Helper-Free AAV Production System?
The Viral Production Cells 2.0 are specifically designed to work with the other components of the AAV MAX Helper-Free AAV Production System to provide maximal adeno-associated virus (AAV) titers. Viral Production Cells could be used but we don't recommend doing this as it will reduce system performance.
What is the highest density that Viral Production Cells 2.0 can be cultured at?
The Viral Production Cells 2.0 achieve maximum cell densities of >1.2 x 10E7 cells/mL in shake flask cultures.
What is the passage stability of the Viral Production Cells 2.0?
We recommend using the Viral Production Cells 2.0 between passages 3 to 25 for adeno-associated virus (AAV) production.
How long do the Viral Production Cells 2.0 require to recover post-thaw before transfection?
We recommend allowing freshly thawed Viral Production Cells 2.0 to recover in culture for at least three passages prior to transfection.
Are the Viral Production Cells 2.0 a clonal cell line?
Yes, the Viral Production Cells 2.0 cells are a clonal cell line derived from single cell cloning of 293-F cells.