TaqMan™ Microbe Detection Assay, pooled - FAQs
TaqMan™ Microbe Detection Assay, pooled - FAQs
View additional product information for TaqMan™ Microbe Detection Assay, pooled - FAQs (A52083)
1 product FAQs found
What are the concentrations of the primers and probes for TaqMan microbial assays?
Our predesigned and custom TaqMan microbial 20X assays have a primer concentration of 900 nM and a probe concentration of 250 nM in the final 1X concentration.
Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.