Oncomine™ Dx Express Test FFPE DNA and RNA Control Kit
Ion Torrent™

Oncomine™ Dx Express Test FFPE DNA and RNA Control Kit

This kit contains DNA, RNA, and no-template controls for use with the Oncomine Dx Express Test.
Have Questions?
Catalog NumberNo. of Reactions
A5216712 Reactions (4 Runs)
Catalog number A52167
Price (MXN)
6,815.43
Each
No. of Reactions:
12 Reactions (4 Runs)
Price (MXN)
6,815.43
Each

The Ion Torrent Oncomine Dx Express Test FFPE DNA and RNA Control Kit contains DNA, RNA, and no-template controls for use with the Oncomine Dx Express Test.

The Oncomine Dx Express Test is an in vitro diagnostic assay that uses targeted next-generation sequencing (NGS) to detect biomarkers across 46 genes. The Oncomine™ Dx Express Test is a qualitative in vitro diagnostic test that uses targeted next generation sequencing (NGS) technology to detect substitutions, insertions, and deletions in 42 genes from DNA, copy number variants (CNVs) in 10 genes from DNA, and fusions or splice variants in 18 genes from RNA isolated from formalin-fixed, paraffin-embedded (FFPE) tumor samples using the Genexus™ Dx Integrated Sequencer.

The test is designed for use with the Genexus Dx System. Its fully automated workflow—from sample preparation to results—can be completed in as little 24 hours, supporting timely decision-making in clinical practice.

For In Vitro Diagnostic Use. Not available in all countries or regions.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Genexus™ Dx Integrated Sequencer
LibrariesAmplicon based
No. of Reactions12 Reactions (4 Runs)
Nucleic Acid TypeDNA, RNA, and No-template
Product LineControl
Product TypeFFPE
Quantity1 Ea.
Unit SizeEach
Contents & Storage
• RNA Control Kit, store at -60 to -90°C
• DNA Control Kit, store at -10 to -30°C
• NTC Control Kit, store at room temperature