Efficient-Pro™ AGT™ Feed 2, 1 L - FAQs
Efficient-Pro™ AGT™ Feed 2, 1 L - FAQs
View additional product information for Efficient-Pro™ AGT™ Feed 2 - FAQs (A5221601, A5221602, A5221603)
5 product FAQs found
Can I use Efficient-Pro AGT Feed 2 with other cell lines besides CHO-S and DG-44 cell line variants?
Efficient-Pro AGT Feed 2 has been designed specifically for CHO-S and DG-44 cells; however, it has been demonstrated to improve performance in other CHO variants and even other parental cell lines.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
I currently supplement my CHO cells with insulin and other growth factors. Should I continue the use of these other additives when using Efficient-Pro AGT Feed 2?
We recommend continuing current supplementation protocols including insulin and other growth factors when using Efficient-Pro AGT Feed 2.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the shelf-life of Efficient-Pro AGT Feed 2?
The shelf-life of Efficient-Pro AGT Feed 2 is 12 months from the date of manufacture. The expiration date is provided on the product label.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How often and at what concentration should I feed my CHO cells with Efficient-Pro AGT Feed 2?
The comprehensive feed protocol for Efficient-Pro AGT Feed 2 is included in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025908_EfficentProAGTFeed2_UG.pdf). We recommend first designing a small-scale experiment using an Ambr15 Microbioreactor or other small vessel with concentrations between 1.5% to 3.0% of Efficient-Pro AGT Feed 2 in 0.5% increments beginning on day 3, with daily feeds through harvest. Once the specific feeding strategy is tailored to your CHO cell system for optimized results, cultures can be scaled up to larger vessels and bioreactors.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
When should I consider using Efficient-Pro AGT Feed 2?
Efficient-Pro AGT Feed 2 is a chemically-defined, animal-origin-free supplement that has been formulated to work specifically with CHO-S and DG-44 cells for production of proteins. Efficient-Pro AGT Feed 2 has been designed to work optimally when paired with Efficient-Pro AGT Medium (https://www.thermofisher.com/order/catalog/product/A5322301). Efficient-Pro AGT Feed 2 has been demonstrated to improve yield and other end-points when used as described in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025908_EfficentProAGTFeed2_UG.pdf).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.