High-Select™ Fe-NTA Magnetic Agarose - FAQs
High-Select™ Fe-NTA Magnetic Agarose - FAQs
View additional product information for High Select™ Phosphopeptide Enrichment Kits & Reagents - FAQs (A52284)
1 product FAQs found
Can I purchase High-Select Fe-NTA Magnetic beads from the High-Select Fe-NTA Magnetic Phosphopeptide Enrichment Kit as a standalone product?
Yes, the standalone beads can be purchased as High-Select Fe-NTA Magnetic Agarose, Cat. No. A52284.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.