Iodoacetamido-LC-Phosphonic Acid (6C-CysPAT)
Thermo Scientific™

Iodoacetamido-LC-Phosphonic Acid (6C-CysPAT)

Iodoacetamido-LC-Phosphonic Acid (6C-CysPAT) is a sulfhydryl-reactive alkylating reagent that contains a phosphonic acid group for enrichment of cysteine-containing peptides for mass spectrometry analysis.
Have Questions?
Catalog NumberQuantity
A5228510 x 3.5 mg
Catalog number A52285
Price (USD)
772.00
Each
Quantity:
10 x 3.5 mg
Price (USD)
772.00
Each
Iodoacetamido-LC-Phosphonic Acid (6C-CysPAT) is a sulfhydryl-reactive alkylating reagent that contains a phosphonic acid group for enrichment of cysteine-containing peptides for mass spectrometry analysis.
  • A sulfhydryl-reactive alkylating reagent that contains a phosphonic acid group for enrichment of cysteine-containing peptides for mass spectrometry analysis
  • Provided in convenient single-use, no-weigh format
  • High-purity, crystalline reagent
  • Compatible with EasyPep chemistry and High Select Phosphopeptide Enrichment kits
Specifications
GradeMS grade
Molecular Weight (g/mol)348.99
Percent Purity>90%
Physical FormSolid
Quantity10 x 3.5 mg
Unit SizeEach

Frequently asked questions (FAQs)

What is the difference between IodoTMT tags and Iodoacetamido-LC-Phosphonic Acid?

Both IodoTMT tags and Iodoacetamido-LC-Phosphonic Acid react with sulfhydryl groups. Iodo TMT tags allow the quantitation of proteins with cysteine amino acids and the oxidative state of sulfhydryl groups, while Iodoacetamido-LC-Phosphonic Acid allow the detection and enrichment of those specific proteins. This is a great tool for mass spectrometry (MS) analysis, and High-Select Fe-NTA Magnetic Phosphopeptide Enrichment Kit (Cat. No. A52283), can be used for the enrichment step.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.