MagMAX™ Wastewater Ultra Nucleic Acid Isolation Kit with Virus Enrichment - FAQs
MagMAX™ Wastewater Ultra Nucleic Acid Isolation Kit with Virus Enrichment - FAQs
View additional product information for MagMAX™ Wastewater Ultra Nucleic Acid Isolation Kit with Virus Enrichment - FAQs (A52610)
4 product FAQs found
Is the purified viral nucleic acid that is obtained using MagMAX Wastewater Ultra Nucleic Acid Isolation Kit, with Virus Enrichment completely inhibitor-free?
The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit, with Virus Enrichment allows elimination of the vast majority of diverse inhibitors of enzymatic reactions, even from the most diluted and challenging samples such as wastewater. In very rare instances, if some inhibitors still remain within the purified nucleic acid, the sample can be diluted 10-20X prior to downstream PCR or other applications.
What starting volume of wastewater can be used with the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit, with Virus Enrichment?
The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit, with Virus Enrichment is compatible with starting volumes of 1-10 mL. If concentration is not needed, or if wastewater has been concentrated using ultracentrifugation or precipitation, use the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit (Cat. No. A52606).
How long is the protocol for the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit, with Virus Enrichment?
The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit, with Virus Enrichment protocol takes 90 mins to concentrate virus and isolate nucleic acid.
Is the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit, with Virus Enrichment automation-compatible?
The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit, with Virus Enrichment is compatible with the KingFisher Flex, APEX, and Duo instruments, and can also be used for manual processing in 24- and 96-well plates.