Slide-A-Lyzer™ G3 Dialysis Cassettes, 10K MWCO, 70 mL, 6 cassettes - FAQs
Slide-A-Lyzer™ G3 Dialysis Cassettes, 10K MWCO, 70 mL, 6 cassettes - FAQs
View additional product information for Slide-A-Lyzer™ G3 Dialysis Cassettes, 10K MWCO - FAQs (A52975, A52974, A52971, A52973, A52972)
1 product FAQs found
What is the surface area of the Slide-A-Lyzer G3 Dialysis Cassettes, 10K MWCO?
The following are the surface areas for the G3 cassettes:
- 3 mL capacity (Cat. No. A52971): 12 cm^2
- 15 mL capacity (Cat. No. A52972): 28 cm^2
- 30 mL capacity (Cat. No. A52973): 50 cm^2
- 70 mL capacity (Cat. No. A52974): 100 cm^2
- 125 mL capacity (Cat. No. A52975): 110 cm^2
Find additional tips, troubleshooting help, and resources within our Protein Biology Support Centers .