QuantStudio™ 7 Pro Dx Real-Time PCR System, 96-well, 0.2 mL, desktop, AB Platinum, IQOQPQ, SmartStart - FAQs
QuantStudio™ 7 Pro Dx Real-Time PCR System, 96-well, 0.2 mL, desktop, AB Platinum, IQOQPQ, SmartStart - FAQs
View additional product information for QuantStudio™ 7 Pro Dx Real-Time PCR System, 96-well, 0.2 mL, desktop, AB Platinum, IQOQPQ, SmartStart - FAQs (A53164)
2 product FAQs found
What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?
Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.
The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
I recently switched the block in my real-time PCR instrument. Now it shows the status as 'not calibrated' for all the calibration plates, even though my calibrations are current. What is causing this issue?
The issue might be due to the block not being properly seated in the instrument. Please follow the steps below:
  1. Remove the block from the instrument.
  2. Reinsert the block into the instrument.
If the problem persists after reseating the block, please contact our Technical Support team for further assistance.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.