Efficient-Pro™ Medium, 20 L - FAQs
Efficient-Pro™ Medium, 20 L - FAQs
View additional product information for Efficient-Pro™ Medium - FAQs (A5322202, A5322201, A5322203)
5 product FAQs found
What is the shelf-life of Efficient-Pro Medium?
The shelf-life of Efficient-Pro Medium is 12 months from the date of manufacture. The expiration date is provided on the product label.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
If my cell line is sensitive to ammonia, can I still use Efficient-Pro Medium?
Yes, however, we recommend reducing the L-glutamine concentration if a decrease in cell viability is observed. The recommended glutamine concentration is 2-8 mM. Another way to reduced ammonia production is to use GlutaMAX Supplement instead of L-glutamine. Addition of Pluronic F-68 is not required.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does Efficient-Pro Medium require supplementation for use?
When using a CHO cell line with a selection system other than the glutamine synthetase (GS) system, we recommend adding 2-8 mM L-glutamine or GlutaMAX Supplement to Efficient-Pro Medium. Additionally, we recommend making sure that glucose levels do not fall below 2 g/L in culture by supplementing with glucose as needed. Also, if clumping becomes an issue, then add anti-clumping agent (ACA) to your cultures at 1% v/v.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the concentration of glucose in Efficient-Pro Medium?
Efficient-Pro Medium contains 4.7 g/L of glucose.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can CHO cells that are created or frozen in other production media be thawed into Efficient-Pro Medium or adapted for growth and production in Efficient-Pro Medium?
Cells that are grown in or adapted to, and frozen in other cell culture media readily adapt to Efficient-Pro Medium. However, if your cells are experiencing a significant lag or delay in growth, please directly adapt the cells into Efficient-Pro Medium and passage them for a minimum of 3 passages, until a consistent doubling time is observed.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.