RapidFinder™ Analysis Software v3.1
Thermo Scientific™
RapidFinder™ Analysis Software v3.1
Optimize your pathogen detection workflow and enhance the downstream decision-making process with this graphical, easy-to-use software.
Have Questions?
Número do catálogoFor Use With (Equipment)
A53289Real Time PCR System
Número do catálogo A53289
Preço (BRL)
-
For Use With (Equipment):
Real Time PCR System
Optimize your pathogen detection workflow and enhance your downstream decision-making process with Thermo Scientific™ RapidFinder™ Analysis Software v3.1. The software simplifies PCR run setup, interpretation of pathogen presence/absence results, and generation of reports including interoperability with LIMS.
The Thermo Scientific™ RapidFinder™ Analysis Software v3.1 is a graphical, easy-to-use tool for food and environmental testing labs to optimize their pathogen detection workflow. The software is required as part of the Applied Biosystems™ QuantStudio™ 5 Food Safety System to run the Thermo Scientific™ SureTect™, Rapid Finder™ and SureCount™ Assays. The RapidFinder™ Analysis Software v3.1:
  • Leads you through the steps to start instrument run
  • Uses pre-set, optimized thermal cycling conditions, and automatically generates the results (presence or absence of the target, CFU/sample in the case of SureCount PCR Assays).
  • Allows you to view results on an easy-to-read screen with color-coded presence/absence calls
  • Offers interrogation of results for the whole plate, grouped by sample, or for individual PCR tubes.
The main features of the RapidFinder Analysis Software include:
  • Fully automated amplification, detection, data collection, and data analysis
  • Quick interpretation of data with flags, notifications, and prompts
  • Detailed report generation for printing or export
  • Interoperability with LIMS or similar data management tool for importing plate set-up and exporting assay results
  • Ability to build database with PCR runs used to search for results and files with user selectable filters
RapidFinder Analysis Software and the QuantStudio 5 Food Safety PCR System with a 96-well, 0.1-mL block configuration are integral parts of the SureTect™, SureCount and RapidFinder™ PCR Assays validated food pathogen testing workflows.
.
For Laboratory Use
For Laboratory Use
Especificações
DescriptionRapidFinder Analysis Software v3.1 downloads
For Use With (Equipment)Real Time PCR System
Unit SizeEach