SAE Administrator Console Software v2.1 - FAQs
SAE Administrator Console Software v2.1 - FAQs
View additional product information for SAE Administrator Console Software v2.1 - FAQs (A53717)
3 product FAQs found
Where can I download the Release Notes for SAE Administrator Console Software v2.1?
Please download this PDF (http://downloads.thermofisher.com/SAE_AdminConsole_v2.1/USB/100108548_SeqStudioFlex_SAE_Admin_Console_Software_Version_2_1_0_Release_Notes.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
I created a user profile in SAE Administrator Console Software v2.0. Can I install it on SAE Administrator Console Software v2.1?
Yes, application profiles created with earlier versions of the SAE software can be used with later versions of the SAE software. However, profiles created with newer SAE servers are not supported by earlier versions.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What applications are compatible with the SAE Administrator Console Software v2.1?
The following applications are compatible with the SAE Administrator Console Software v2.1:
- SeqStudio Flex Series Instrument Software v1.0 and above
- Design and Analysis Software v2.5.0 and above
- QuantStudio 6/7 Pro Version 1.4.0 Instrument Software and above
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.