How many reactions come in a single order of the Absolute Q Custom FAM dPCR Assay (Cat. No. A53734)?
The Absolute Q Custom FAM dPCR Assay (Cat. No. A53734) comes as a 350 µL volume and can be used in 700 reactions (0.5 µL/reaction).