Genexus™ Dx Barcodes 1-32 HD - FAQs
Genexus™ Dx Barcodes 1-32 HD - FAQs
View additional product information for Genexus™ Dx Barcodes 1-32 HD - FAQs (A54104)
3 product FAQs found
What is included in Genexus Dx Barcodes 1-32 HD (Cat. No. A54104)?
Genexus Dx Barcodes 1-32 HD provides one plate filled with 32 lyophilized, single-use, dual barcodes (Barcodes HD 1-32) and covered with a pierceable foil seal, eliminating the risk of cross contamination between wells. Each plate supports up to 32 reactions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Will the Genexus Dx Software notify me if the barcodes had been previously used?
When necessary, the software will alert the user during the run plan setup that not enough barcodes are available.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long can the Genexus Dx Barcodes remain on the Genexus Dx platform?
The Genexus Dx Barcode plates can remain on the instrument deck until all barcodes are used. Once you remove the barcode plate, you cannot put it back on to use again as the instrument has no way of tracking barcode usage.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.