Genexus™ Dx Barcodes 1-32 HD
Ion Torrent™

Genexus™ Dx Barcodes 1-32 HD

The Ion Torrent Genexus Dx Barcodes 1-32 HD kit provides one plate filled with 32 dried-down, single-use barcodes and covered with a pierceable foil seal.
Have Questions?
Catalog NumberNo. of Reactions
A5410432 Reactions
Catalog number A54104
Price (USD)
-
No. of Reactions:
32 Reactions

The Ion Torrent Genexus Dx Barcodes 1-32 HD kit provides one plate filled with 32 dried-down, single-use barcodes and covered with a pierceable foil seal. The barcodes can be flexibly used in increments of 1 to 32, until all barcodes are exhausted.

These barcodes are used with the Genexus Dx Integrated Sequencer, which is an in vitro diagnostic (IVD) medical device intended to be used by trained personnel in a professional laboratory environment for automated, qualitative targeted sequencing of human DNA or RNA extracted from formalin-fixed, paraffin-embedded (FFPE) tumor tissue samples or plasma samples.

For In Vitro Diagnostic Use. Not available in all countries or regions.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Genexus™ Dx Integrated Sequencer
LibrariesTargeted Sequencing Library
No. of Reactions32 Reactions
Nucleic Acid TypeDNA, RNA, and cfTNA* (*Not available in the U.S.)
Quantity1 Ea.
Sequencing TypeGenome, DNA, and RNA Sequencing
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

What is included in Genexus Dx Barcodes 1-32 HD (Cat. No. A54104)?

Genexus Dx Barcodes 1-32 HD provides one plate filled with 32 lyophilized, single-use, dual barcodes (Barcodes HD 1-32) and covered with a pierceable foil seal, eliminating the risk of cross contamination between wells. Each plate supports up to 32 reactions.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Will the Genexus Dx Software notify me if the barcodes had been previously used?

When necessary, the software will alert the user during the run plan setup that not enough barcodes are available.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How long can the Genexus Dx Barcodes remain on the Genexus Dx platform?

The Genexus Dx Barcode plates can remain on the instrument deck until all barcodes are used. Once you remove the barcode plate, you cannot put it back on to use again as the instrument has no way of tracking barcode usage.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.