Genexus™ Dx Pipette Tips - FAQs
Genexus™ Dx Pipette Tips - FAQs
View additional product information for Genexus™ Dx Pipette Tips - FAQs (A54105)
1 product FAQs found
How many pipette tips do I get with Genexus Dx Pipette Tips (Cat. No. A54105)?
12 racks of pipette tips are provided, containing 96 tips per rack. The pipette tips can be stored at room temperature.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.