CTS™ HiFi Cas9 Protein
Gibco™

CTS™ HiFi Cas9 Protein

Gibco CTS HiFi Cas9 Protein is our GMP-manufactured high-fidelity CRISPR-Cas9 protein, engineered to deliver reduced off-target effects while maintaining highRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A542232.5 mg
A542245 mg
Catalog number A54223
Price (USD)
-
Quantity:
2.5 mg

Gibco CTS HiFi Cas9 Protein is our GMP-manufactured high-fidelity CRISPR-Cas9 protein, engineered to deliver reduced off-target effects while maintaining high on-target activity. This protein is designed and manufactured in compliance with the principles of GMP and to meet requirements of the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products. It is provided in a high-concentration and large-scale format for process development and clinical research applications.

Features of CTS HiFi Cas9 Protein include:
• Exceptional off-target profile—over 70% reduction in off-target effects in primary immune cells, compared to CTS TrueCut Cas9 (wild type) at tested loci
• Maintain on-target activity—achieve comparable on-target activity to CTS TrueCut Cas9 (wild type) in primary immune cells at tested loci
• Stringent manufacturing process—manufactured to meet the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products including USP , Ph.Eur. 5.2.12, and ISO 20399 following the principles of 21 CFR Part 820 in an FDA-registered manufacturing site
• Extensive safety testing—includes sterility, endotoxin, mycoplasma, residual nucleic acid, and protein testing
• Traceability documentation—COA, COO, Drug Master files, and Regulatory Support files* are available

CTS HiFi Cas9 Protein is a Streptococcus pyogenes high-fidelity Cas9 protein, produced by recombinant DNA technology in E. coli for genome editing with CRISPR technology. Cas9 protein forms a very stable RNP complex with the guide RNA (gRNA) component of the CRISPR-Cas9 system. Incorporation of NLS aids delivery to the nucleus, increasing the editing efficiency and on-target cleavage.

If you would like to evaluate our HiFi Cas9 protein or conduct small-scale research experiments, we recommend using our research-grade TrueCut HiFi Cas9 Protein.


*Country-specific Regulatory Support File process will be initiated upon request and will require additional documentation time.

For Research Use or Non-Commercial Manufacturing of Cell-Based Products for Clinical Research. Caution: Not intended for direct administration into humans or animals.
Specifications
Concentration10 mg/mL
Expression SystemE. coli
Manufacturing QualityISO 13485
Product TypeCas9 Protein
Quantity2.5 mg
Research CategoryGenome Editing
Shelf Life36 months from date of manufacture
Shipping ConditionDry Ice
FormLiquid
Unit SizeEach
Contents & Storage
Store at –20°C.

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

Who can I contact for outlicensing questions regarding Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221)?

The Limited Use Label Licenses are available on the individual product pages. For additional questions, please contact outlicensing@thermofisher.com.

Where do I find the traceability documentation for Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221)?

To request traceability documentation for Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221), please follow the instructions at the following link: https://www.thermofisher.com/us/en/home/clinical/cell-gene-therapy/cell-therapy/cell-therapy-systems/cell-therapy-systems-support.html

How is Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) different from Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221)?

Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221) is a Streptococcus pyogenes Cas9 wild-type protein, produced by recombinant DNA technology in E. coli and Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) is a Streptococcus pyogenes high-fidelity CRISPR Cas9 protein, produced by recombinant DNA technology in E. coli. Both proteins are designed and manufactured in compliance with GMP.

How is Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) different from Invitrogen TrueCut HiFi Cas9 Protein (Cat. No. A50574, A50575)?

Both Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Invitrogen TrueCut HiFi Cas9 Protein (Cat. No. A50574, A50575) are Streptococcus pyogenes high-fidelity CRISPR Cas9 proteins, produced by recombinant DNA technology in E. coli. Both proteins have the same sequence with added nuclear localization signal. Gibco CTS HiFi Cas9 Protein is manufactured to meet the standards for “Ancillary Materials for Cell, Gene, and Tissue-Based Products” whereas Invitrogen TrueCut HiFI Cas9 Protein is a research-grade product.