RapidINTEL™ Plus 50 Sample Cartridge Kit
Applied Biosystems™

RapidINTEL™ Plus 50 Sample Cartridge Kit

The Applied Biosystems RapidINTEL Plus 50 Sample Cartridge Kit is used with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to generate lab-quality forensic DNA profiles.
Have Questions?
Catalog NumberQuantity
A543381 Kit (50 Cartridges)
Catalog number A54338
Price (USD)
8,650.00
Each
Quantity:
1 Kit (50 Cartridges)
Price (USD)
8,650.00
Each

The Applied Biosystems RapidINTEL Plus 50 Sample Cartridge Kit is used with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to generate lab-quality forensic DNA profiles. The DNA profiles generated from RapidINTEL Plus sample cartridges can be compared to DNA profiles obtained from crime scene evidence to link one crime to another or to DNA profiles from reference samples to link a crime to an individual.

This kit contains 50 RapidINTEL Plus Sample Cartridges, 2 RapidINTEL Plus Positive Control Cartridges, and 2 RapidINTEL Plus Negative Control Cartridges, each of which is an individually-wrapped, single-use, disposable cartridge that contains reagents for developing a forensic DNA profile from a single sample in 90 minutes.

The RapidINTEL 'Plus' sample cartridge is an upgraded cartridge that provides more complete, more consistent data from crime scene samples. In addition, internal quality control (IQC) markers and quantification markers have been added to assist users with identification of low quantity, degraded, and inhibited samples.

To run reference samples on the Applied Biosystems RapidHIT ID System, see our ACE GlobalFiler Sample Cartridge Kit (Cat. No. A41831 or 400114RH).

Required for use

  • Applied Biosystems RapidHIT ID System (Cat. No. A41810)
  • RapidHIT ID Instrument Software v2.0 or higher
  • RapidHIT ID Primary Cartridge (Cat. No. A53084)

For Research Use Only.

Specifications
For Use With (Application)Human Identification
For Use With (Equipment)RapidHIT™ ID
No. of Samples50
Quantity1 Kit (50 Cartridges)
Sample TypeCrime scene samples
Shelf Life12 Months
FormatKit
Unit SizeEach
Contents & Storage
Contains 50 sample cartridges, 2 positive control cartridges, and 2 negative control cartridges.