RapidINTEL™ Plus Positive Control Cartridge
Applied Biosystems™

RapidINTEL™ Plus Positive Control Cartridge

The Applied Biosystems RapidINTEL Plus Positive Control Cartridge is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to verify instrument and/or reagent performance.
Have Questions?
Catalog NumberQuantity
A543421 Cartridge
Catalog number A54342
Price (USD)
298.00
Each
Quantity:
1 Cartridge
Price (USD)
298.00
Each

The Applied Biosystems RapidINTEL Plus Positive Control Cartridge is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to verify instrument and/or reagent performance. Each cartridge is individually wrapped and single use/disposable.

Required for use

  • Applied Biosystems RapidHIT ID System (Cat. No. A41810)
  • RapidHIT ID Instrument Software v2.0 or higher
  • RapidHIT ID Primary Cartridge (Cat. No. A53084)

For Research Use Only.

Specifications
For Use With (Application)Human Identification
For Use With (Equipment)RapidHIT™ ID
Quantity1 Cartridge
Shelf Life12 Months
FormatCartridge
Unit SizeEach