Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
CTS™ StemFlex™ Medium
Gibco™

CTS™ StemFlex™ Medium

CTS StemFlex Medium is a xeno-free medium designed for robust expansion of feeder-free pluripotent stem cells (PSCs) and supports stressful applications in translational and clinical applications.
Have Questions?
Catalog NumberQuantity
A5465001500 mL
Catalog number A5465001
Price (USD)
672.00
500 mL
Quantity:
500 mL
Price (USD)
672.00
500 mL
CTS (Cell Therapy Systems) StemFlex Medium is a xeno-free medium designed for robust expansion of feeder-free pluripotent stem cells (PSCs). The optimized formulation supports stressful applications for translational and clinical research and offers a flexible feeding schedule. PSCs expanded in CTS StemFlex Medium maintain stable karyotype, pluripotency, and tri-lineage differentiation potential across multiple passages and cell lines.

CTS StemFlex Medium:

  • Delivers reliable performance for gene editing and single cell passaging.
  • Offers a flexible feeding schedule.
  • Supports the transition from other adherent culture media.
  • Facilitates regulatory filings with GMP manufacturing, traceability of raw materials, and regulatory documentation.

Robust recovery and clonal expansion

CTS StemFlex Medium promotes consistent growth across multiple passages and supports high viability post single-cell passaging. The medium delivers efficient recovery following electroporation and achieves editing efficiencies similar to research-use-only (RUO) StemFlex Medium. CTS StemFlex Medium supports clonal expansion while maintaining pluripotency and normal karyotype of edited clones.

Workflow flexibility

CTS StemFlex Medium supports a flexible feeding schedule, including weekend-free feeding, and is compatible with different matrices and passaging reagents.

Simple transition

Induced pluripotent stem cells (iPSCs) and embryonic stem cells (ESCs) grown in other adherent culture media can be transitioned to CTS StemFlex Medium within a few passages without the need for extensive optimization. Additionally, cells cultured in CTS StemFlex Medium can be adapted to CTS StemScale PSC Suspension Medium, helping achieve consistent fold expansion per passage in suspension culture.

GMP manufactured

Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820, helping ensure consistent quality. Our FDA-registered manufacturing site is ISO 13485-certified. Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available for CTS StemFlex Medium.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell LineEmbryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Cell TypeEmbryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
ConcentrationCTS StemFlex Basal Medium (1X), CTS StemFlex Supplement (10X)
Expression SystemNone
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatBottle
FormulationLiquid
Manufacturing QualityISO13485, MDSAP, FDA-registered, 21 CFR820
Product TypeCell Culture Medium
Quantity500 mL
Shelf Life12 Months from Date of Manufacture
Shipping ConditionCTS StemFlex Basal Medium - ambient, CTS StemFlex Supplement - dry ice
ClassificationXeno-free
Culture TypeAdherent Cell Culture, Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic), Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesNone
Without AdditivesNo Phenol Red
Unit Size500 mL
Contents & Storage
450 mL CTS StemFlex Basal Medium: store at 2°C to 8°C, protect from light. 50 mL CTS StemFlex Supplement: store at -5°C to -20°C, protect from light.

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.