Thermo Scientific™ 96-well Pierceable Seals
Thermo Scientific™
Thermo Scientific™ 96-well Pierceable Seals
Eliminate lysis tube capping and uncapping steps with the specifically designed Thermo Scientific 96-well Pierceable Seals in the SureTect workflow.
Have Questions?
Catalog NumberMaterial
A55331Polyester
Catalog number A55331
Price (MXN)
-
Material:
Polyester
Speed up the lysis and pipetting steps during the SureTect™ workflow using the Thermo Scientific™ 96-well Pierceable Seals.
Designed specifically for use with the SureTect™ assays the 96-well Pierceable Seals offer the ability to quickly and securely seal and pipette from the lysis tubes, reducing cross contamination and eliminating the requirement for capping and uncapping steps to enable a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.
Use the Thermo Scientific 96-well Pierceable Seals to streamline your SureTect PCR Assay workflow, quickly and easily sealing lysis plates and pipette from the SureTect lysis tubes directly through the seal, reducing cross-contamination and eliminating the need for capping and uncapping to enable a fast and easy to use workflow.
The high integrity sealing materials feature a strong adhesive providing secure plate sealing during cycling and sample storage.
Printed well positions and orientation markings enable you to position and identify samples
Strip perforations for ease of separation and use regardless of your sample throughput.
Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.
Specifications
Description96-well Pierceable Seals
For Use With (Application)SureTect PCR Assay workflow
ColorClear
MaterialPolyester
PierceableYes
Unit SizeEach