HID NIMBUS™ QNA System
Applied Biosystems™

HID NIMBUS™ QNA System

The Applied Biosystems HID NIMBUS QNA System is an easy-to-use, compact automated instrument that can set up a DNA quantification plate, perform normalization, and set up an STR amplification plate for your forensic needs.
Have Questions?
Catalog NumberQuantity
A55771Each
Catalog number A55771
Price (USD)
-
Quantity:
Each

The Applied Biosystems HID NIMBUS QNA System is an easy-to-use, compact automated instrument that can set up a DNA quantification plate, perform normalization, and set up an STR amplification plate for your forensic needs.

This automated solution is optimized for use with Applied Biosystems forensic human identification (HID) quantification and STR amplification kits, which have been validated according to the Scientific Working Group for DNA Analysis Methods (SWGDAM, December 2016).

Quantification setup
Cover more steps in the casework workflow with the optimized scripting and hands-free automated quantification setup. The HID NIMBUS QNA system can prepare samples for quantification using the Quantifiler Trio Automated DNA Quantification Kit either on a QuantStudio 5 or a 7500 real-time PCR system.

  • Improved amplification efficiency
  • Fewer off-scale, over-amplified, or under-amplified samples
  • Reduced need for downstream re-analysis helps provide long-term cost savings
  • Key sample information to enable forensic workflow decisions
  • Conservation of sample through targeted downstream assay decisions

Normalization and amplification setup
The HID NIMBUS QNA System can remove the manual normalization process by automatically calculating dilution protocol and normalizing DNA samples. Add automated amplification setup, and analysts to help reduce hands on time and experience support, reliable, and accurate results from a variety of forensically relevant sample types.

Software
The internally designed software includes visualization guides for reagent positioning, guidance mode for step-by-step setup, short tutorial videos, and images of the work deck setup. Users can initiate purification, quantification setup, and amplification setup in just a few minutes.

  • Easily select workflows, kits, and input/output labware
  • Select from commonly used plate map templates or customize your own (and add to library)
  • Standard curves: 5 pt, 6 pt, or virtual
  • Visual cues and guidance mode to enable accurate and easy setup

For Criminal Justice and Forensic Use Only.

Specifications
Dimensions (D x W x H)53.5 x 27.9 x 35 in. (135.9 x 70.9 x 88.9 cm)
Electrical RequirementsPower input: 100-240 VAC, 50-60 Hz, 5 A
For Use With (Equipment)Automated Quantification Plate set up, Normalization and Amplification Plate set up
Frequency50-60 Hz
Height (English)35 in. (door closed), 51.2 in (door open)
Height (Metric)88.9 cm (door closed), 130.0 cm (door open)
IncludesDell Laptop
QuantityEach
SoftwareHID NIMBUS System Software (included)
Temperature (Metric) Operating0°C to 40°C
Throughput1 to 96 Samples
Voltage100/240 V
Warranty1 Year
Weight (English)247 lb.
Weight (Metric)112.1 kg
TypeAccessioning and Sample Transfer Instrument
Unit SizeEach